Learn More
Medchemexpress LLC Tinidazole (Standard) | 19387-91-8 | 99.9% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0177R
Encompass_Preferred
Tinidazole (Standard) is an analytical standard and an orally available antibacterial agent. It is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa, intended for research and analytical applications.
  • Analytical standard for Tinidazole
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Selective activity against anaerobic bacteria and protozoa
Catalog No. 50-004-07440