Learn More
Medchemexpress LLC Tinidazole (Standard) | 19387-91-8 | 99.95% | 50 MG

Supplier: Medchemexpress LLC HYB0177R50MG
Tinidazole (Standard) is the analytical standard of Tinidazole, an orally available antibacterial agent and a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments.
- Analytical standard of Tinidazole
- Orally available antibacterial agent
- Selective activity against anaerobic bacteria and protozoa
- Suitable for qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS
- Purity of 99.95%
- Off-white solid appearance
- Molecular formula: C8H13N3O4S
- Molecular weight: 247.27
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.