Learn More
Sigma Aldrich Fine Chemicals Biosciences TinII chloride 99.99 trace10G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 20472210G
Encompass
Tin(II) chloride (Stannous chloride SnCl2) is widely employed as reducing reagent. It plays significant role in diverse fields of science. Since it exhibits superior catalytic action in molten state it is employed for the hydroliquefaction of coals. It is a promising raw material for the chemical vapour deposition (CVD) of semiconducting layers. It can be prepared by reacting its dihydrate form with acetic anhydride. It exhibits reducing properties in acidic media. It participates in the reduction of the following compoundsaromatic nitro compoundsnitriles cyanosilyl ethersorganic azidesOn mixing sodium cyanoborohydride with SnCl2 in a 21 ratio a reducing reagent mixture is obtained. This reagent is useful for the reduction of tertiary allyl and benzyl halides.
Catalog No. 50-289-7770