Learn More
Sigma Aldrich Fine Chemicals Biosciences TinII chloride dihydrate p100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 31669100G
Encompass
Tin(II) chloride dihydrate is the hydrated salt of tin(II). Its crystals are monoclinic and their structure has been investigated by single crystal X-ray diffraction method. Cell parameters were reported to be a9.313 b7.250 c8.970A… 11455AE and Z4.
Catalog No. 50-289-9721