Learn More
Sigma Aldrich Fine Chemicals Biosciences TinII chloride free-flowin500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 908924500G
Encompass
SnCl2 dihydrate reacts with acetic anhydride to yield anhydrous SnCl2. SnCl2 exhibits reducing properties in acidic media. SnCl2 shows superior catalytic action in its molten state. It is a promising raw material for the chemical vapour deposition (CVD) of semiconducting layers. SnCl2 participates in the reduction of aromatic nitro compounds nitriles cyanosilyl ethers and organic azides.
Catalog No. 50-290-3349