Learn More
Sigma Aldrich Fine Chemicals Biosciences TinII oxide 60 micron part500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 244643500G
Encompass
Tin(II) oxide (SnO) is a metal oxide with wide applications in superconductivity photovoltaics optoelectronics as an anode coating substrate Schottky diode and catalyst.
Catalog No. 50-289-8894