Learn More
Medchemexpress LLC Tiopronin-13C,d3 | 1189695-13-3 | 95.3% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0373S5MG
Encompass_Preferred
Tiopronin-13C,d3 is deuterium labeled Tiopronin. It is intended for research use only and is not sold to patients.
  • This compound can be used as a tracer
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06844