Learn More
Medchemexpress LLC Tiopronin-13c,d3 | 1189695-13-3 | ≥98% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0373S1MG
Encompass_Preferred
Tiopronin-13C,d3 is a deuterium labeled form of Tiopronin. Tiopronin is known as a diffusible antioxidant, an antidote for heavy metal poisoning, and a radioprotective agent. This compound can help control the rate of cystine precipitation and excretion, showing potential for use in treating conditions like cystinuria, rheumatoid arthritis, and various hepatic disorders.
  • Deuterium labeled Tiopronin
  • Functions as a diffusible antioxidant
  • Acts as an antidote to heavy metal poisoning
  • Serves as a radioprotective agent
  • Shows potential for cystinuria, rheumatoid arthritis, and hepatic disorders
Catalog No. 50-004-06646