Learn More
Medchemexpress LLC HY-101232 5mg Medchemexpress, Tiotidine CAS:69014-14-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012325MG
Encompass_Preferred
Medchemexpress, HY-101232 5mg Tiotidine CAS:69014-14-8 Tiotidine (ICI 125211) is a potent and selective antagonist of histamine H2-receptor (pA2=7.3-7.8 for guinea-pig right atrium). Tiotidine has low affinity for both the H1 and the H3 receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7814