Learn More
Apexbio Technology LLC Tiotropium Bromide, 100mg. Cas: 136310-93-5 MFCD: MFCD00867027. MAChR M antagonist
SDP
Supplier:  Apexbio Technology LLC A3874100
Encompass_Preferred
Tiotropium Bromide is a muscarinic acetylcholine receptor (mAChR M) antagonist that blocks the binding of the acetylcholine ligand and subsequent opening of the ligand-gated ion channel. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4846