Learn More
Apexbio Technology LLC Tiotropium Bromide hydrate 139404-48-1 5mg
SDP
Supplier:  Apexbio Technology LLC B16195
Encompass_Preferred
Tiotropium Bromide hydrate is a small-molecule antagonist targeting muscarinic acetylcholine receptors (mAChRs) It is designed to competitively inhibit mAChRs thereby modulating airway smooth muscle contraction and mediating bronchodilation Tiotropium Bromide hydrate exerts its biological activity primarily through competitive antagonism at mAChRs resulting in relaxation of airway smooth muscle Based on these pharmacological properties Tiotropium Bromide hydrate holds research potential in respiratory pharmacology particularly for investigating mechanistic pathways in chronic obstructive pulmonary disease (COPD) receptor-ligand interactions and pulmonary pharmacodynamics associated with airway obstruction
Catalog No. 50-000-88278