Learn More
Selleck Chemical LLC Tiotropium Bromide hydrate S2547-5mg
SDP
Supplier:  Selleck Chemical LLC S25475MG
Encompass_Preferred
Tiotropium Bromide hydrate (BA 679BR) is a monohydrate of tiotropium bromide (Spiriva Tiova BA 679BR tiopropium) that is an anticholinergic and bronchodilator and a muscarinic receptor antagonist
Catalog No. 50-311-1881