Learn More
Labcon North America ZAP™ 100uL Extended Length Aerosol Filter Pipet Tips, in Racks, Sterile
Supplier:  Labcon North America 10989650089
Encompass_Preferred
Tip, Pipet; Labcon; ZAP™ Aerosol Filter; FlexTop™ and Extended Length; Fits most popular 100uL Pipettors; Low mounting and ejection forces less than 3kg; Sterile; Patented robotically inserted endotoxin free filter; Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Validated Sterilization (ISO 11137) for use in Research and Medical labs; Refillable autoclavable racks; 96 tips/rack; 6 racks/pack; 8 packs/case; 83.8mm H.; Made in USA
Catalog No. 50-101-582