Learn More
Waters Corp Tip Insertion System, for Andrew+ Robot, 96x OptiFit Tips
Supplier:  Waters Corp 186009612
Encompass_Preferred
  • Capable of holding a removable rack of 96x Sartorius OptiFit tips.
  • Supports tips for volumes ranging from 0.2μL to 1200μL.
  • Force-dispensing design ensures even tip insertion across all channels of 8-channel pipettes.
  • Machine-readable consumable holders.
  • Creates an interface between different consumable types and the Andrew+ pipetting robot for seamless automation.
  • Equipped with QR codes for critical labware information.
  • Features ARUCO codes for spatial positioning.
Catalog No. NC2131705