Learn More
Molecular Devices Molecular Devices Pipette Tips, 384-Well, Black, 50 Racks/Case
Supplier:  Molecular Devices 90000911
Encompass_Preferred
These FLIPR 384-well black pipette tips are designed for reliable automated dispensing with FLIPR integrated pipettors and compatible plate readers. They are certified RNase-/DNase-free and supplied in multi-rack packaging to support high-throughput workflows.
  • Compatible with integrated pipettors and select plate readers.
  • 384-well SBS format for high-density assays.
  • Black tips to reduce assay cross-talk in fluorescence applications.
  • Certified RNase-/DNase-free and nonpyrogenic.
  • Nonsterile, packaged 50 racks per case.
Catalog No. 50-000-14399