Learn More
Labcon North America APT™ 50uL Automation Pipet Tips for Workstations, in Refills
Supplier:  Labcon North America 80102000209
Encompass_Preferred
Tip, Pipet; Automated Workstation; Labcon; APT™; 50uL; Fits 50uL Beckman 96 pipettors; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Autoclavable; Magenta color; 2688 tips per low waste refill; 8 refills per case; 48.3mm. H; Made in USA
Catalog No. 50-101-602