Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Dumont Tweezer Replacement Tip DIRPY1, Delrin, 45 mm OAL
SDP

Supplier:  Electron Microscopy Sciences 7672315

Encompass_Preferred

Replacement delrin tip for DIRPY1 replaceable tip tweezers.
Material: Delrin
OAL: 45 mm
Tip thickness: 1 mm

Catalog No. 50-193-1119


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.