Learn More
Medchemexpress LLC Tipepidine hydrochloride | 1449686-84-3 | 100.0% | 1 ML

Supplier: Medchemexpress LLC HY121685A10MM 1ML
Tipepidine hydrochloride is a research-grade small molecule that acts as an IDA(GIRK) potassium channel inhibitor and is used to study dopaminergic neuron activity and related neuropharmacology. It is supplied as a ready-to-use 10 mM solution in DMSO for in vitro assays.
- Research reagent for potassium channel (IDA/GIRK) inhibition studies.
- High purity, suitable for biochemical and cell-based assays.
- Molecular formula C15H18ClNS2; molecular weight 311.89 g/mol.
- Supplied as a 10 mM solution in DMSO for convenient dosing in assays.
- Soluble in DMSO and water according to product specifications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.