Learn More
Medchemexpress LLC Tipiracil | 183204-74-2 | 99.98% | C9H11ClN4O2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HYA0063A200MG
Encompass_Preferred
Tipiracil is a thymidine phosphorylase (TPase) inhibitor intended for research use only. It increases trifluridine exposure by inhibiting its metabolism by thymidine phosphorylase. The combination of Trifluridine and Tipiracil is a new oral treatment for metastatic colorectal cancer.
  • Thymidine phosphorylase (TPase) inhibitor
  • Increases trifluridine exposure
  • Prevents degradation of FTD
  • New oral treatment for metastatic colorectal cancer
Catalog No. 50-004-32139