Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.9% | C9H12Cl2N4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0063100MG
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) used for cancer research. It exhibits antiangiogenic effects and inhibits the proliferation of endothelial cells. This compound is also a potent inhibitor of thymidine phosphorylase, an enzyme that degrades FTD. It is for research use only.
  • Thymidine phosphorylase inhibitor (TPI)
  • Exhibits antiangiogenic effects
  • Inhibits the proliferation of endothelial cells
  • Potent inhibitor of thymidine phosphorylase
Catalog No. 50-004-32317