Learn More
Medchemexpress LLC Tipiracil hydrochloride | 183204-72-0 | 99.9% | C9H12Cl2N4O2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HYA0063200MG
Encompass_Preferred
Tipiracil hydrochloride is a thymidine phosphorylase inhibitor (TPI) that is used for cancer research. It exhibits an antiangiogenic effect and inhibits the proliferation of endothelial cells. Tipiracil is a potent inhibitor of thymidine phosphorylase, the enzyme that degrades FTD.
  • Thymidine phosphorylase inhibitor (TPI)
  • Antiangiogenic effect
  • Inhibits proliferation of endothelial cells
  • Potent inhibitor of thymidine phosphorylase
Catalog No. 50-004-32123