Learn More
Medchemexpress LLC Tiplaxtinin | 393105-53-8 | C24H16F3NO4 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY1525310MM/1ML
Encompass_Preferred
Tiplaxtinin is a selective and orally efficacious inhibitor of plasminogen activator inhibitor-1 (PAI-1) with an IC50 of 2.7 μM.
  • Selective and orally efficacious inhibitor of PAI-1.
  • IC50 of 2.7 μM for PAI-1.
  • Inhibits cellular proliferation in T24 cells and UM-UC-14 cells.
  • Reduces thrombus weight in a rat model of thrombosis.
  • Reduces subcutaneous tumor growth in human bladder cancer and human cervical cancer xenografts.
Catalog No. 50-003-72913