Learn More
Selleck Chemical LLC Tiplaxtinin (PAI-039) 10mg 393105-53-8
SDP
Supplier:  Selleck Chemical LLC S792210MG
Encompass_Preferred
Tiplaxtinin(PAI-039) is an orally efficacious and selective plasminogen activator inhibitor-1 (PAI-1) inhibitor with IC50 of 2.7 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5418