Learn More
USA Scientific Inc 1000 ul XL TipOne RPT filter tip, sterile, case of 4 packs
Supplier:  USA Scientific Inc 11821880
Encompass
TipOne RPT 1000 ul ultra low retention filter tip in racks, 4-unit case, sterile, 40 racks of 96 tips (total 3840 tips)
Catalog No. NC2002910