Learn More
Medchemexpress LLC Tipranavir-d7 | 174484-41-4 | C31H26D7F3N2O5S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15148S11MG
Encompass_Preferred
Tipranavir-d7 is a deuterated form of Tipranavir, an inhibitor of HIV-1 protease that also inhibits SARS-CoV-2 3CLpro activity. It is designed for research applications, particularly as a tracer or internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Its stable heavy isotope labeling allows for studies on pharmacokinetic and metabolic profiles.
  • Inhibits enzymatic activity and dimerization of HIV-1 protease.
  • Potent activity against multi-protease inhibitor-resistant HIV-1 isolates.
  • Inhibits SARS-CoV-2 3CLpro activity.
  • Can be used as a tracer in research.
  • Functions as an internal standard for quantitative analysis.
  • Stable isotope labeling aids in drug development process.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-72311