Learn More
CELLTREAT Scientific Products 10µL Extended Length Low Retention Reload System, Non-sterile
SDP
Supplier:  CELLTREAT Scientific Products 229053
Encompass_Preferred
Reload System offers clear window for easy identification of color-coded wafersSupport wafers have large grasping tabs for easy removal from Reload System and RackSecure locking tab keeps reload system closed and prevents tumbling tipsReduce plastic waste and save time by refilling empty racksCELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing processes results in superior low retentionPipette Tips have smooth, hydrophobic surfaces to ensure accurate pipettingMultiple Pipette Tip sizes are available, including extended length tips (10μL and 1000μL) to allow better access to the bottom of test tubes, reagent bottles, flasks and other vesselsPipette Tips are universal fit and compatible with major pipettesFine tip for improved accuracy and precisionMolded volume reference markings offer a quick double check opportunity
Catalog No. NC2545522