Learn More
NanoScreen LLC TIPS 30UL 384 50/CS BS PE COMP
Supplier:  NanoScreen LLC TIPP30384SB
Encompass_Preferred
Pipette tips, Robotic. PerkinElmer P30, 384 tips. Sterile, Aerosol Barrier. For PE/Packard 384 dispense heads found on MiniTrak, Janus and EP3. Tips are 100% inspected for quality. Guaranteed 1536 compatible. Case=50 racks=19200 tips
Catalog No. 50-921-312