Learn More
Medchemexpress LLC Tirabrutinib (hydrochloride) | 1439901-97-9 | MFCD28386296 | 99.7% | 490.94 g/mol | C25H23ClN6O3 | 10 MG

Supplier: Medchemexpress LLC HY15771A10MG
Tirabrutinib hydrochloride is the hydrochloride salt of tirabrutinib, a selective, orally active Bruton's tyrosine kinase (BTK) inhibitor supplied as a research-grade powder for preclinical and biochemical studies.
- Hydrochloride salt form, suitable for in vitro and in vivo research.
- High purity (99.7%).
- Chemical formula C25H23ClN6O3; molecular weight 490.94 g/mol.
- CAS number 1439901-97-9.
- Supplied as a powder; typical pack size 10 MG.
- Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
- Function: selective BTK inhibitor for signaling and oncology research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.