Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tirbanibulin dihydrochloride | 1038395-65-1 | 98.0% | 504.45 g/mol | C26H31Cl2N3O3 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY10340A100MG

Encompass_Preferred

Tirbanibulin dihydrochloride is the dihydrochloride salt form of tirbanibulin supplied as an analytical research standard for in vitro testing and analytical applications. It functions as a Src inhibitor with reported GI50 values in the low nanomolar range in cancer cell lines.

  • Analytical standard suitable for research use and assay development.
  • Dihydrochloride salt form; molecular weight 504.45 g/mol.
  • CAS number 1038395-65-1 for unambiguous identification.
  • Purity (HPLC) 98.02% as stated on the certificate of analysis.
  • Available in small pack sizes including 100 mg for analytical workflows.

Catalog No. 50-002-44767


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.