Learn More
Medchemexpress LLC Tirbanibulin dihydrochloride | 1038395-65-1 | MFCD18633218 | 98.0% | 504.45 g·mol⁻¹ | C26H31Cl2N3O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10340A10MM 1ML
Encompass_Preferred
Tirbanibulin dihydrochloride is a research-grade Src inhibitor provided as the dihydrochloride salt for in vitro cellular and biochemical studies. The material is characterized by a molecular formula of C26H31Cl2N3O3, a molecular weight of 504.45 g·mol⁻¹, and HPLC-confirmed purity of approximately 98%.
  • Research-grade Src inhibitor (dihydrochloride salt)
  • Intended for in vitro cellular and biochemical assays
  • HPLC purity approximately 98.0%
  • Molecular formula C26H31Cl2N3O3; molecular weight 504.45 g·mol⁻¹
  • Supplied as a solid (powder) in common pack sizes; solution formats may be supplier-specific
Catalog No. 50-002-46488