Learn More
Medchemexpress LLC Tirofiban (hydrochloride monohydrate) | 150915-40-5 | MFCD07368623 | 99.9% | 495.07 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17369
Encompass_Preferred
Tirofiban hydrochloride monohydrate is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist that inhibits fibrinogen binding to this receptor and has antithrombotic activity. It induces proliferation and migration on endothelial cells by inducing the production of VEGF. It can significantly reduce myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area. This product is for research use only.
  • Increases proliferation of HAEC cells.
  • Closes the scratch of HUVECs migration.
  • Induces production of VEGF, stimulating proliferation of endothelial cells.
  • Increases contraction force and improves heart function in rats after acute myocardial infarction.
  • Enhances eNOS activity and reduces no-reflow area after reperfusion.
  • Shows anticoagulant effect in a crush injury model in rats.
Catalog No. 50-003-84587