Learn More
KRISHGEN BIOSYSTEMS Enzyme Immunoassay for Quantitative Estimation of Tirzepatide in human serum and plasma. Assay Range: 50 - 4000 ng/ml.
Supplier:  KRISHGEN BIOSYSTEMS KOD1027
Encompass_Preferred
Pack Size: 1 x 96 wells. Introduction: This novel Tirzepatide ELISA is optimised to detect tirzepatide in human samples. If you would like to use animal samples, please contact us. Validated against ICH M10 guidelines for bioassays. Introduction: Tirzepatide, a novel dual GIP and GLP-1 receptor agonist, is an antidiabetic medication used for the treatment of type 2 diabetes and for weight loss. A linear polypeptide of 39 amino acids, it has been chemically modified by lipidation to improve its uptake into cells and its stability to metabolism. It is an analogue of gastric inhibitory polypeptide (GIP), a human hormone that stimulates the release of insulin from the pancreas. ELISA Features include a pre-coated plate with break-apart wells, standardisation and Lot to Lot Consistency with Precision CV <15%.
Catalog No. NC2747160