Learn More
Medchemexpress LLC Tisolagiline methylsulfate | 1894207-45-4 | 99.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY127109B
Encompass_Preferred
Tisolagiline (KDS2010) methylsulfate is a selective, reversible, and orally active inhibitor for MAO-B with an IC50 of 8 nM. It demonstrates neuroprotective and anti-neuroinflammatory activities.
  • Selective, reversible, and orally active MAO-B inhibitor
  • IC50 of 8 nM
  • Demonstrates neuroprotective activities
  • Demonstrates anti-neuroinflammatory activities
Catalog No. 50-003-38704