Learn More
Ihc World LLC TISSUE FLOTATION BATH
SDP
Supplier:  Ihc World LLC TFB200101
Encompass_Preferred
ype Tissue Flotation BathModel TFB200101Temperature Range 0-100°C (1°C increment)Control 5" Color Touch ScreenSlide Placement Area Room for 8+ SlidesGlass Basin Dimensions 210mm x 168mm x 55mmUnit Dimensions 327mm x 340mm x 166mmTemperature Sensor InternalControl Functions Light / Timer / Auto ON&OFF / Light / Heater / AlarmElectrical AC120V/60Hz | 200WHeating Time 20 minutes to heat to 54°CTemperature Accuracy +/- 1°CRelative Humidity <80%Suitable Ambient Temperature 5-30°CLight Color WhiteMaterials of Construction Epoxy Painted Steel with PTFE coated Slide Drying Area
Catalog No. NC3449953