Learn More
Chemglass Life Sciences Tissue Grinder, Dounce, 1mL Working Capacity, 11mm Dia Pestle, 48mm Chamber Lgth
SDP
Supplier:  Chemglass Life Sciences CLS500301
Encompass_Preferred
Tissue Grinder, Dounce, 1mL Working Capacity, 11mm Dia Pestle, 48mm Chamber Lgth. The Dounce tissue grinder is used by moving the pestle up and down and is designed for studies where the nucleus has to remain intact Supplied with two pestles that are used in the same tube, the large clearance pestle is for the initial sample reduction while the small clearance pestle is used to form the final homogenate.
Catalog No. 50-153-2380