Learn More
BioSpec Products Tissue-Tearor with 7 mm Probe
SDP
Supplier:  BioSpec Products 98537007
Encompass_Preferred
The Tissue-Tearor is a rotor/stator type tissue homogenizer which can rapidly homogenize, disrupt, and emulsify samples in 0.5 - 1000 ml of liquid.  Complete homogenization of tissues (muscle, liver, breast tissue, etc.) is usually achieved in ten to thirty seconds. Little, if any, heat is produced during the process. Practical Working Range (ml) .5-50 Probe Diameter (mm) 7 Probe Length (cm) 8.3
Catalog No. NC0433502
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.