Learn More
Andwin Scientific TISSUE TEK MARKING PENCIL
SDP
Supplier:  Andwin Scientific M732110
Encompass_Preferred
The Tissue-Tek Marking Pencils are ideal for writing on cassettes and rings. The soft-lead pencils feature smudge-resistant markings. The markings can endure several types of liquids, such as chloroform,acetone,alcohol and molten paraffin. This product is sold in a pack of 12.
Catalog No. NC9668506