Learn More
TA Instruments Titanium, 50 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Supplier:  TA Instruments 402634.901
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Titanium Quick Change Plate (50 mm) is compatible with Advanced Peltier Plates and ARES-G2 APS systems. Durable and corrosion resistant, it ensures precise placement, reproducible measurements, and reliable sample interaction.
Catalog No. 50-005-80803