Learn More
Medchemexpress LLC Titanium dioxide | 13463-67-7 | MFCD00011269 | 99.0% | 79.87 g/mol | TiO2 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW051271
Encompass_Preferred
Titanium dioxide (TiO2) is a white inorganic powder widely used as a pigment, photocatalyst, and pharmaceutical excipient. It is supplied as a fine powder with typical purity around 99.0% and a molar mass of 79.87 g/mol. Common applications include coatings, pigments, catalyst studies, and formulation use where a stable, inert white filler or coating agent is required. Documentation such as a product datasheet, safety data sheet, and certificate of analysis are provided.
  • High purity (99.0%) suitable for research and formulation.
  • Fine white powder ideal for coatings and pigments.
  • Acts as a photosensitizer and photocatalyst in photochemical studies.
  • Usable as a pharmaceutical excipient for coating and flow aid.
  • Available in laboratory pack sizes, including 250 g.
  • Supplied with datasheet, safety data sheet, and certificate of analysis.
Catalog No. 50-002-09596