Learn More
Strem, An Ascensus Company CAS# 546-68-9. 250g. Titanium(IV) i-propoxide, min. 98%. MFCD00008871
SDP
Supplier:  Strem, An Ascensus Company 932216250G
Encompass
CAS# 546-68-9. 250g. Titanium(IV) i-propoxide, min. 98%. MFCD00008871. Molecular Weight: 284.25. Molecular Formula: Ti[OCH(CH3)2]4. Color/form: colorless to pale yellow liq. Strem# 93-2216. http://www.strem.com/catalog/v/93-2216/
Catalog No. 50-901-14441