Learn More
Selleck Chemical LLC Titanium tetrafluoride S4872-1g
SDP
Supplier:  Selleck Chemical LLC S48721G
Encompass_Preferred
Titanium tetrafluoride is an inorganic compound that is an effective but instable caries preventive agent
Catalog No. 50-311-1040