Learn More
Sigma Aldrich Fine Chemicals Biosciences TitaniumIV oxide anatase n500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 637254
Encompass
Titanium (IV) oxide nanopowder is widely used in skin products additives and foods. Nano titania has been proved to be the most active photocatalytic semiconductor under ultraviolet radiation. Titanium isopropoxide titanium (IV) butoxide has been used as precursor materials to synthesize nano-titania.
Catalog No. 50-290-6712