Learn More
Titration Solvent, ASTM D664, D974, D3242, Reagents
Titration Solvent, ASTM D664, D974, D3242, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$206.50 - $695.50
Specifications
| Chemical Name or Material | Titration Solvent, ASTM D664, D974, D3242 |
|---|---|
| Color | Colorless |
| CAS | 108-88-3 , 67-63-0 , 7732-18-5 |
| Boiling Point | 90°C |
| Grade | Reagent |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
01-333-439
|
Reagents Holdings Llc
CT12500020E6 |
20 L | Steel Pail |
Each for $695.50
|
|
||||
|
01-333-438
|
Reagents Holdings Llc
CT1250004C |
4 L | Glass Bottle |
Each for $206.50
|
|
||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.