Learn More
Titration Solvent, ASTM D664, D974, D3242, Reagents
Specifications
Specifications
| Chemical Name or Material | Titration Solvent, ASTM D664, D974, D3242 |
| Color | Colorless |
| CAS | 108-88-3, 67-63-0, 7732-18-5 |
| Boiling Point | 90°C |
| Grade | Reagent |
| Packaging | Steel Pail |
| Specific Gravity | 0.8245 |
| Quantity | 20 L |
| Sterility | Non-sterile |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.