Learn More
Medchemexpress LLC Tivantinib | 905854-02-6 | 99.6% | C23H19N3O2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY50686200MG
Encompass_Preferred
Tivantinib is a highly selective, low-molecular-weight, and orally available c-Met tyrosine kinase inhibitor. It selectively inhibits c-Met activity in both cell-free and cell-based assays, leading to a dose-dependent loss of proliferative capacity or caspase-dependent apoptosis in c-Met-expressing cancer cell lines.
  • Highly selective c-Met tyrosine kinase inhibitor
  • Inhibits c-Met activity in cell-free and cell-based assays
  • Causes dose-dependent loss of proliferative capacity or apoptosis in c-Met-expressing cancer cells
  • Orally available
  • Inhibits constitutive c-Met phosphorylation
  • Inhibits HGF-induced c-Met phosphorylation
Catalog No. 50-003-58746