Learn More
Apexbio Technology LLC Tivantinib (ARQ 197), 5mg. Cas: 905854-02-6 MFCD: MFCD11977597. C-Met inhibitor,non-ATP-competitive
SDP
Supplier:  Apexbio Technology LLC A83255
Encompass_Preferred
Tivantinib (ARQ 197) is an oral, nonadenosine triphosphate-competitive, selective, small-molecule met proto-oncogene (c-MET) inhibitor. The calculated inhibitory constant (Ki) for tivantinib to inhibit recombinant human c-MET was approximately 355 nmol/L. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5110