Learn More
Medchemexpress LLC Tivozanib | 475108-18-0 | 454.86 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10977100MG
Encompass_Preferred
Tivozanib (Standard) is the analytical standard of Tivozanib, intended for research and analytical applications. It acts as a selective, orally active inhibitor for vascular endothelial growth factor receptor (VEGFR)-1, 2, and 3. This compound is suitable for use in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard of Tivozanib
  • Intended for research and analytical applications
  • Selective, orally active inhibitor for VEGFR-1, 2, and 3
  • Exhibits antitumor efficacy
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-003-44540