Learn More
Medchemexpress LLC Tixocortol-21-pivalate | 55560-96-8 | 96.8% | 462.64 g·mol⁻¹ | C26H38O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1303655MG
Encompass_Preferred
Tixocortol pivalate is a synthetic corticosteroid used in research for its anti-inflammatory properties. It is provided as a purified solid for in vitro pharmacology and allergy/contact-sensitivity studies and should be stored under controlled refrigerated or frozen conditions.
Catalog No. 50-002-62875