Learn More
Apexbio Technology LLC Tizanidine HCl 64461-82-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13685.1
Encompass_Preferred
Tizanidine hydrochloride (CAS 64461-82-1) is a small molecule that functions as an agonist at 2-adrenergic receptors By activating these presynaptic receptors it suppresses the release of neurotransmitters from central noradrenergic neurons thereby modulating synaptic transmission in the central nervous system This mechanism makes Tizanidine HCl a valuable tool for investigating 2-adrenergic signaling pathways neurotransmitter regulation and CNS synaptic function in biomedical research settings
Catalog No. 50-000-91365