Learn More
Medchemexpress LLC Tizanidine hydrochloride | 64461-82-1 | MFCD00798231 | >98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0194A100MG
Encompass_Preferred
Tizanidine hydrochloride is a skeletal muscle relaxant and an orally effective central α2-adrenoceptor agonist (IC50 = 6.9 nmol). It primarily exerts muscle relaxation effects by inhibiting the release of excitatory amino acids (glutamate and aspartate) from the presynaptic terminals of spinal cord interneurons. It possesses anti-injury activity and can inhibit gastrointestinal transport.
  • Inhibits the proliferation, migration, and invasion of lung cancer cells.
  • Induces cell apoptosis by upregulating Nischarin.
  • Inhibits AKT and Wnt3a/β-catenin signaling pathways.
  • Used to treat spasticity caused by diseases such as multiple sclerosis, stroke, and spinal cord injury.
Catalog No. 50-004-07032